COM448613153 Combi-Blocks, Inc.

[2-Methoxy-4-(piperidin-1-yl)pyrimidin-5-yl]boronic acid

  • MDL number MFCD27935639
  • Molecular Weight : 237.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0436

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0436
Availability details for
Quantity
  • Availability
  • Add to Cart