COM448613156 Combi-Blocks, Inc.

1-(2-Fluorophenyl)-1h-pyrazole-4-boronic acid

  • MDL number MFCD21099664
  • Cas Number : 1374984-33-4
  • Molecular Weight : 206

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0439

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0439
Availability details for
Quantity
  • Availability
  • Add to Cart