COM448613157 Combi-Blocks, Inc.

[4-(3-Methylpyrazol-1-yl)phenyl]boronic acid

  • MDL number MFCD27935642
  • Cas Number : 2225172-13-2
  • Molecular Weight : 202.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0440

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0440
Availability details for
Quantity
  • Availability
  • Add to Cart