COM448613163 Combi-Blocks, Inc.

{4-[(Dimethylcarbamothioyl)oxy]-3-fluorophenyl}boronic acid

  • MDL number MFCD27935648
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0446

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0446
Availability details for
Quantity
  • Availability
  • Add to Cart