COM448613165 Combi-Blocks, Inc.

[2-Cyano-3-(pyrrolidin-1-yl)phenyl]boronic acid

  • MDL number MFCD27935649
  • Molecular Weight : 216.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0451
Availability details for
Quantity
  • Availability
  • Add to Cart