COM448613170 Combi-Blocks, Inc.

[3-(2-Methyl-1,3-thiazol-4-yl)phenyl]boronic acid

  • MDL number MFCD26937712
  • Molecular Weight : 219.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0457
Availability details for
Quantity
  • Availability
  • Add to Cart