COM448613171 Combi-Blocks, Inc.

[3-Cyano-4-(trifluoromethoxy)phenyl]boronic acid

  • MDL number MFCD27935655
  • Cas Number : 2268800-99-1
  • Molecular Weight : 230.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0459

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0459
Availability details for
Quantity
  • Availability
  • Add to Cart