COM448613174 Combi-Blocks, Inc.

[4-(1-Cyanocyclopropyl)-2-methylphenyl]boronic acid

  • MDL number MFCD27935658
  • Molecular Weight : 201.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0462

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0462
Availability details for
Quantity
  • Availability
  • Add to Cart