COM448613175 Combi-Blocks, Inc.

4-(1-Cyanocyclopropyl)-3-fluorophenylboronic acid

  • MDL number MFCD27935659
  • Cas Number : 2377611-05-5
  • Molecular Weight : 205.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0463

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0463
Availability details for
Quantity
  • Availability
  • Add to Cart