COM448613176 Combi-Blocks, Inc.

{8-Cyano-7-methoxyimidazo[1,2-a]pyridin-3-yl}boronic acid

  • MDL number MFCD27935660
  • Molecular Weight : 217.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0464

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0464
Availability details for
Quantity
  • Availability
  • Add to Cart