COM448613193 Combi-Blocks, Inc.

(5-Fluoro-2-methanesulfonylphenyl)boronic acid

  • MDL number MFCD27935676
  • Cas Number : 2377607-65-1
  • Molecular Weight : 218.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0481
Availability details for
Quantity
  • Availability
  • Add to Cart