COM448613196 Combi-Blocks, Inc.

[2-Cyano-3-(piperidin-1-yl)phenyl]boronic acid

  • MDL number MFCD27935679
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0484

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0484
Availability details for
Quantity
  • Availability
  • Add to Cart