COM448613198 Combi-Blocks, Inc.

[3-Fluoro-2-(pyrazol-1-ylmethyl)phenyl]boronic acid

  • MDL number MFCD27935681
  • Molecular Weight : 220.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0486

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0486
Availability details for
Quantity
  • Availability
  • Add to Cart