COM448613199 Combi-Blocks, Inc.

{[2-Fluoro-3-(trifluoromethoxy)phenyl]methyl}boronic acid

  • MDL number MFCD27935682
  • Molecular Weight : 237.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0488

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0488
Availability details for
Quantity
  • Availability
  • Add to Cart