COM448613201 Combi-Blocks, Inc.

(2-Methanesulfonyl-5-methylphenyl)boronic acid

  • MDL number MFCD27935685
  • Cas Number : 1447763-90-7
  • Molecular Weight : 214.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0491

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0491
Availability details for
Quantity
  • Availability
  • Add to Cart