COM448613203 Combi-Blocks, Inc.

[2-Chloro-4-(1-cyanocyclopropyl)phenyl]boronic acid

  • MDL number MFCD27935687
  • Cas Number : 2377609-59-9
  • Molecular Weight : 221.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0493

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0493
Availability details for
Quantity
  • Availability
  • Add to Cart