COM448613219 Combi-Blocks, Inc.

(6-Fluoro-1-isopropyl-1,3-benzodiazol-5-yl)boronic acid

  • MDL number MFCD27935703
  • Molecular Weight : 222.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0510

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0510
Availability details for
Quantity
  • Availability
  • Add to Cart