COM448613222 Combi-Blocks, Inc.

[3,5-Difluoro-4-(2,2,2-trifluoroethoxy)phenyl]boronic acid

  • MDL number MFCD27935706
  • Cas Number : 160483-71-6
  • Molecular Weight : 255.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0514

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0514
Availability details for
Quantity
  • Availability
  • Add to Cart