COM448613225 Combi-Blocks, Inc.

3-Cyano-4-pyrrolidinophenylboronic acid

  • MDL number MFCD27931158
  • Cas Number : 876918-90-0
  • Molecular Weight : 216.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0521

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0521
Availability details for
Quantity
  • Availability
  • Add to Cart