COM448613242 Combi-Blocks, Inc.

3-(1,1-Dioxido-2-isothiazolidinyl)phenylboronic acid

  • MDL number MFCD22573107
  • Cas Number : 1176093-78-9
  • Molecular Weight : 241.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0543

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0543
Availability details for
Quantity
  • Availability
  • Add to Cart