COM448613245 Combi-Blocks, Inc.

[4-(3-Methoxyphenyl)pyrimidin-5-yl]boronic acid

  • MDL number MFCD27935725
  • Molecular Weight : 230.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0546

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0546
Availability details for
Quantity
  • Availability
  • Add to Cart