COM448613247 Combi-Blocks, Inc.

[3-(5-Butyl-1,3,4-oxadiazol-2-yl)phenyl]boronic acid

  • MDL number MFCD27935727
  • Molecular Weight : 246.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0548

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0548
Availability details for
Quantity
  • Availability
  • Add to Cart