COM448613251 Combi-Blocks, Inc.

{4-Chloro-2-[2-(morpholin-4-yl)ethoxy]phenyl}boronic acid

  • MDL number MFCD27935731
  • Cas Number : 2377605-74-6
  • Molecular Weight : 285.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0555

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0555
Availability details for
Quantity
  • Availability
  • Add to Cart