COM448613252 Combi-Blocks, Inc.

[2-(4-Ethylpiperazin-1-yl)-5-methylpyridin-3-yl]boronic acid

  • MDL number MFCD27935733
  • Molecular Weight : 249.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0557

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0557
Availability details for
Quantity
  • Availability
  • Add to Cart