COM448613255 Combi-Blocks, Inc.

{4-Fluoro-3-[(4-methylpiperazin-1-yl)methyl]phenyl}boronic acid

  • MDL number MFCD27935736
  • Cas Number : 1704063-92-2
  • Molecular Weight : 252.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0560

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0560
Availability details for
Quantity
  • Availability
  • Add to Cart