COM448613263 Combi-Blocks, Inc.

{3-[(1E)-(tert-Butylimino)methyl]-4-fluorophenyl}boronic acid

  • MDL number MFCD27935743
  • Molecular Weight : 223.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0568

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0568
Availability details for
Quantity
  • Availability
  • Add to Cart