COM448613266 Combi-Blocks, Inc.

(7-Chloro-2,8-dimethylquinolin-4-yl)boronic acid

  • MDL number MFCD27935746
  • Molecular Weight : 235.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0571

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0571
Availability details for
Quantity
  • Availability
  • Add to Cart