COM448613268 Combi-Blocks, Inc.

5-(1,1-Dioxo-1,2-thiazolidin-2-yl)-2-methylphenylboronic acid

  • MDL number MFCD27935748
  • Molecular Weight : 255.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0573

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0573
Availability details for
Quantity
  • Availability
  • Add to Cart