COM448613276 Combi-Blocks, Inc.

(1-Butyl-6-fluoro-2-methyl-1,3-benzodiazol-5-yl)boronic acid

  • MDL number MFCD27935757
  • Molecular Weight : 250.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0582
Availability details for
Quantity
  • Availability
  • Add to Cart