COM448613277 Combi-Blocks, Inc.

{[3-(Benzyloxy)phenyl]methyl}boronic acid

  • MDL number MFCD27935758
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0583

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0583
Availability details for
Quantity
  • Availability
  • Add to Cart