COM448613286 Combi-Blocks, Inc.

[1-(3-Chloropyridin-2-yl)-3,5-dimethylpyrazol-4-yl]boronic acid

  • MDL number MFCD27935767
  • Molecular Weight : 251.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0592

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0592
Availability details for
Quantity
  • Availability
  • Add to Cart