COM448613297 Combi-Blocks, Inc.

{3-Cyclohexyl-7-methylimidazo[4,5-b]pyridin-6-yl}boronic acid

  • MDL number MFCD27935777
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0603

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0603
Availability details for
Quantity
  • Availability
  • Add to Cart