COM448613299 Combi-Blocks, Inc.

(6,7-Difluoro-1-isopropyl-1,3-benzodiazol-5-yl)boronic acid

  • MDL number MFCD27935779
  • Molecular Weight : 240.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0605

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0605
Availability details for
Quantity
  • Availability
  • Add to Cart