COM448613304 Combi-Blocks, Inc.

[4-(Dimethoxymethyl)-2-(trifluoromethyl)phenyl]boronic acid

  • MDL number MFCD27935784
  • Molecular Weight : 264.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0610

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0610
Availability details for
Quantity
  • Availability
  • Add to Cart