COM448613305 Combi-Blocks, Inc.

[6-(Trifluoromethoxy)quinolin-8-yl]boronic acid

  • MDL number MFCD27935785
  • Cas Number : 1089179-11-2
  • Molecular Weight : 257.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0611

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0611
Availability details for
Quantity
  • Availability
  • Add to Cart