COM448613307 Combi-Blocks, Inc.

6-(Dihydroxyboranyl)isoquinolin-2-ium-2-olate

  • MDL number MFCD27935787
  • Cas Number : 1690326-28-3
  • Molecular Weight : 189.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0613

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0613
Availability details for
Quantity
  • Availability
  • Add to Cart