COM448613308 Combi-Blocks, Inc.

(6-Cyano-2-methyl-3-propyl-1,3-benzodiazol-4-yl)boronic acid

  • MDL number MFCD27935788
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0615

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0615
Availability details for
Quantity
  • Availability
  • Add to Cart