COM448613324 Combi-Blocks, Inc.

[4-(5-tert-Butyl-1,2,4-oxadiazol-3-yl)phenyl]boronic acid

  • MDL number MFCD27935804
  • Molecular Weight : 246.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0633

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0633
Availability details for
Quantity
  • Availability
  • Add to Cart