COM448613325 Combi-Blocks, Inc.

[3-(Diethylsulfamoyl)-5-methylphenyl]boronic acid

  • MDL number MFCD27935805
  • Molecular Weight : 271.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0634

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0634
Availability details for
Quantity
  • Availability
  • Add to Cart