COM448613326 Combi-Blocks, Inc.

[3-Methyl-5-(pyrrolidine-1-sulfonyl)phenyl]boronic acid

  • MDL number MFCD27935806
  • Cas Number : 2377611-76-0
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0635

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0635
Availability details for
Quantity
  • Availability
  • Add to Cart