COM448613327 Combi-Blocks, Inc.

1-(3-Trifluoromethylphenyl)-1h-pyrazole-4-boronic acid

  • MDL number MFCD21099661
  • Cas Number : 2225178-71-0
  • Molecular Weight : 256

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0636

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0636
Availability details for
Quantity
  • Availability
  • Add to Cart