COM448613329 Combi-Blocks, Inc.

{[4-(2-Cyanophenyl)phenyl]methyl}boronic acid

  • MDL number MFCD27935808
  • Molecular Weight : 237.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0638

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0638
Availability details for
Quantity
  • Availability
  • Add to Cart