COM448613331 Combi-Blocks, Inc.

[5-Methyl-2-(4-propylpiperazin-1-yl)pyridin-3-yl]boronic acid hydrochloride

  • MDL number MFCD27935809
  • Molecular Weight : 299.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0640

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0640
Availability details for
Quantity
  • Availability
  • Add to Cart