COM448613333 Combi-Blocks, Inc.

(4-{6-Methylimidazo[1,2-a]pyridin-2-yl}phenyl)boronic acid

  • MDL number MFCD27935811
  • Cas Number : 2377608-29-0
  • Molecular Weight : 252.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0642

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0642
Availability details for
Quantity
  • Availability
  • Add to Cart