COM448613334 Combi-Blocks, Inc.

[6-(5-Methyl-1,3,4-oxadiazol-2-yl)naphthalen-2-yl]boronic acid

  • MDL number MFCD27935812
  • Molecular Weight : 254.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0643

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0643
Availability details for
Quantity
  • Availability
  • Add to Cart