COM448613336 Combi-Blocks, Inc.

[2-(Pyrazole-1-sulfonyl)phenyl]boronic acid

  • MDL number MFCD27935814
  • Molecular Weight : 252.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0645

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0645
Availability details for
Quantity
  • Availability
  • Add to Cart