COM448613345 Combi-Blocks, Inc.

[6-(Trifluoromethoxy)quinolin-8-yl]boronic acid hydrochloride

  • MDL number MFCD27935823
  • Molecular Weight : 293.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0655

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0655
Availability details for
Quantity
  • Availability
  • Add to Cart