COM448613359 Combi-Blocks, Inc.

[3-Methanesulfonyl-4-(trifluoromethoxy)phenyl]boronic acid

  • MDL number MFCD27935837
  • Molecular Weight : 284.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0672

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0672
Availability details for
Quantity
  • Availability
  • Add to Cart