COM448613361 Combi-Blocks, Inc.

[6-(5-Propyl-1,3,4-oxadiazol-2-yl)naphthalen-2-yl]boronic acid

  • MDL number MFCD27935839
  • Molecular Weight : 282.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0674
Availability details for
Quantity
  • Availability
  • Add to Cart