COM448613363 Combi-Blocks, Inc.

{3-[5-(Trichloromethyl)-1,3,4-oxadiazol-2-yl]phenyl}boronic acid

  • MDL number MFCD27935841
  • Molecular Weight : 307.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0676

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0676
Availability details for
Quantity
  • Availability
  • Add to Cart